Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acinus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500216
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.Specifications
Acinus | |
Polyclonal | |
Western Blot 1:500 -1:1000 | |
Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
Rabbit | |
IgG purified | |
RUO | |
Primary | |
Reacts with the N terminus of Acinus. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
ACIN1 | |
C-terminus of Acinus (proprietary peptide). | |
0.1 mg | |
Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
22985 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction