Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acinus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Acinus |
---|---|
Dilution | Western Blot 1:500 -1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.Specifications
Acinus | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
PBS with 0.02% Sodium Azide | |
22985 | |
C-terminus of Acinus (proprietary peptide). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500 -1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
ACIN1 | |
IgG | |
IgG purified | |
Reacts with the N terminus of Acinus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title