Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACKR2 Mouse anti-Human, Clone: 1A3A2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ACKR2 |
---|---|
Clone | 1A3A2 |
Concentration | 0.83 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6205
![]() ![]() |
Proteintech Group Inc
600461IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.Specifications
ACKR2 | |
0.83 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACKR2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6 | |
ACKR2 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
1A3A2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
O00590 | |
1238 | |
ACKR2 Fusion Protein Ag1941 | |
Primary | |
-20°C | |
ACKR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title