Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AcmNPV gp64 Protein Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB293200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293200 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293200 Supplier Novus Biologicals Supplier No. NBP326087AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AcmNPV gp64 Protein Polyclonal antibody specifically detects AcmNPV gp64 Protein in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen AcmNPV gp64 Protein
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant AcmNPV gp64 Protein (Accession#: NP_054158.1, Met1-Thr481)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.