Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aconitase 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$416.50 - $682.00
Specifications
Antigen | Aconitase 1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Aconitase 1 Polyclonal specifically detects Aconitase 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Aconitase 1 | |
Polyclonal | |
Rabbit | |
metabolism | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
48 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:YERIHRSNLVGMGVIPLEYLPGENADALGLTGQERYTIIIPENLKPQMKVQVKLDTGKTFQAVMRFDTDVELTYFLNGGILNYMIRKMAK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Aconitase, aconitase 1, soluble, aconitate hydratase, Citrate hydro-lyase, cytoplasmic aconitate hydratase, EC 4.2.1, EC 4.2.1.3, Ferritin repressor protein, IREB1IREBP1, IREBP, IRE-BP 1, Iron regulatory protein 1, iron-responsive element binding protein 1, Iron-responsive element-binding protein 1, IRP1ACONS | |
ACO1 | |
IgG | |
Affinity Purified | |
Specificity of human Aconitase 1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title