Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aconitate hydratase B Antibody (02), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB353572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353572 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353572 Supplier Novus Biologicals Supplier No. NBP306089MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aconitate hydratase B Monoclonal antibody specifically detects Aconitate hydratase B in E. coli samples. It is validated for ELISA

Specifications

Antigen Aconitate hydratase B
Applications ELISA
Classification Monoclonal
Clone 02
Conjugate mFluor Violet 450 SE
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias acnB, hypothetical protein, yacI, yacJ
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Aconitate hydratase B (Uniprot#: P36683; Met1-Val865).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.