Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT11 Antibody (J4B2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP14289646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14289646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP14289646 Supplier Novus Biologicals Supplier No. NBP104289JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACOT11 Monoclonal specifically detects ACOT11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ACOT11
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone J4B2
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acyl-CoA thioester hydrolase 11, acyl-CoA thioesterase 11BFIT1, Adipose-associated thioesterase, BFIT2, BFITDKFZp667O1916, Brown fat-inducible thioesterase, EC 3.1.2.-, EC 3.1.2.1, KIAA0707acyl-coenzyme A thioesterase 11, STARD14, START domain containing 14, THEAStAR-related lipid transfer (START) domain containing 14, THEM1, thioesterase superfamily member 1, thioesterase, adipose associated
Gene Symbols ACOT11
Host Species Mouse
Immunogen Recombinant human ACOT11 (19-250aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26027
Test Specificity Anti-human ACOT11 mAb, clone J4B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ACOT11 protein.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.