Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT12 Antibody (OTI1A11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272389G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272389G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272389G Supplier Novus Biologicals Supplier No. NBP272389G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACOT12 Monoclonal specifically detects ACOT12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen ACOT12
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1A11
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acyl-CoA thioester hydrolase 12, acyl-CoA thioesterase 12EC 3.1.2.1, acyl-coenzyme A thioesterase 12, Cach, CACH1, CACH-1, Cytoplasmic acetyl-CoA hydrolase 1, cytosolic acetyl-CoA hydrolase, hCACH-1, StARD15, STARD15MGC105114, StAR-related lipid transfer (START) domain containing 15, START domain-containing protein 15, THEAL
Gene Symbols ACOT12
Host Species Mouse
Immunogen Full length recombinant protein of human ACOT12 (NP_570123) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 134526.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.