Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532152
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA532152 100 μL
1 options

Catalog No. PIPA532152

Supplier: Invitrogen™ PA532152

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (86%), Rat (82%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hydrolyzes acetyl-CoA to acetate and CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ACOT12
Gene Accession No. Q8WYK0
Gene Alias 1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL
Gene Symbols ACOT12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ACOT12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.