Learn More
Invitrogen™ ACOT2 Polyclonal Antibody

Description
ACOT2 Polyclonal Antibody detects endogenous levels of ACOT2 protein.
Specifications
Specifications
| Antigen | ACOT2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ACOT2 |
| Gene Accession No. | P49753 |
| Gene Alias | AA571646; Acot2; Acyl coenzyme A thioester hydrolase; acyl-CoA thioesterase 2; Acyl-coenzyme A thioester hydrolase 2a; acyl-coenzyme A thioesterase 2, mitochondrial; ARTISt/p43; CTE Ia; CTE1A; CTE-Ia; long-chain acyl-CoA thioesterase 2; mitochondrial acyl-CoA thioesterase 1; Mte1; MTE-I; peroxisomal long-chain acyl-coA thioesterase 2; PTE2; PTE2A; Very-long-chain acyl-CoA thioesterase; ZAP128 |
| Gene Symbols | ACOT2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.