Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT7 Antibody (1D5), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N222569APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N222569APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N222569APC7 Supplier Novus Biologicals Supplier No. NBP222569APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACOT7 Monoclonal specifically detects ACOT7 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ACOT7
Applications Flow Cytometry
Classification Monoclonal
Clone 1D5
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ACH1, acyl-CoA thioesterase 7CTE-IIa, BACHEC 3.1.2.2, brain acyl CoA hydrolase, Brain acyl-CoA hydrolase, CTE-IILACH, cytosolic acyl coenzyme A thioester hydrolase, hBACH, long chain, Long chain acyl-CoA thioester hydrolase
Gene Symbols ACOT7
Host Species Mouse
Immunogen Recombinant human ACOT7 (1-370aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11332.0
Test Specificity Anti-human ACOT7 mAb, clone 1D5, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ACOT7 protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.