Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT8 Antibody (4D10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB160881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160881 Supplier Novus Biologicals Supplier No. NBP259419AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACOT8 Monoclonal antibody specifically detects ACOT8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ACOT8
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 4D10
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III
Host Species Mouse
Immunogen Recombinant human ACOT8 (1-319aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10005
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.