Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | ACP1 |
---|---|
Concentration | 0.52 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118924
|
Invitrogen™
PA5118924 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122294
|
Invitrogen™
PA5122294 |
20 μL |
Each for $209.50
|
|
|||||
Description
ACP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.Specifications
ACP1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
ACP1 | |
4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334) | |
ACP1 | |
Primary | |
Antigen affinity chromatography |
0.52 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P41498 | |
24161 | |
Recombinant protein Acid Phosphatase 1. The antigen corresponds to amino acid range 1-158 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title