Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP162491
Description
ACP2 Polyclonal specifically detects ACP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACP2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
acid phosphatase 2, lysosomal, EC 3.1.3.2, LAP, lysosomal acid phosphatase | |
Rabbit | |
45 kDa | |
100 μL | |
Protein Phosphatase | |
53 | |
Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
P11117 | |
ACP2 | |
Synthetic peptides corresponding to ACP2(acid phosphatase 2, lysosomal) The peptide sequence was selected from the middle region of ACP2 (NP_001601). Peptide sequence VPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANET. | |
Affinity purified | |
RUO | |
Primary | |
Bovine: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction