Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACP2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ACP2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP162491
SDP
View Documents
Novus Biologicals
NBP162491
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ACP2 Polyclonal specifically detects ACP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ACP2
Polyclonal
Rabbit
Protein Phosphatase
acid phosphatase 2, lysosomal, EC 3.1.3.2, LAP, lysosomal acid phosphatase
ACP2
IgG
45 kDa
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
P11117
53
Synthetic peptides corresponding to ACP2(acid phosphatase 2, lysosomal) The peptide sequence was selected from the middle region of ACP2 (NP_001601). Peptide sequence VPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANET.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.