Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein Antibody (10A10), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N259359PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N259359PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N259359PEC5 Supplier Novus Biologicals Supplier No. NBP259359PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Flow Cytometry.

Specifications

Antigen Acrolein
Applications Flow Cytometry
Classification Monoclonal
Clone 10A10
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Host Species Mouse
Immunogen Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.