Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrolein diethyl acetal, 96%
Click to view available options
Quantity:
10 g
50 g
Description
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 0.852 |
Boiling Point | 124°C to 125°C |
Flash Point | 13°C (55°F) |
Refractive Index | 1.401 |
Quantity | 10 g |
UN Number | UN2374 |
Beilstein | 1701567 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 130.19 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H225-H319 Highly flammable liquid and vapour. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 3,3-DIETHOXYPROPENE |
EINECSNumber | 221-276-0 |
RTECSNumber | AS1370000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction