Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ BKV Controls
GSA_VA

Catalog No. p-7000144
Click to view available options
Level:
High
Low

Catalog No. 23-279-140

Supplier: Thermo Scientific™ 960051

Only null left
Add to Cart
Add to Cart

Identify increases in random or systematic error of nucleic acid test procedures for BKV DNA with these controls, which are for In Vitro Diagnostic Use.

Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of BKV DNA with the Thermo Scientific™ AcroMetrix™ BKV Controls. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for In Vitro Diagnostic Use.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix BKV High Control
Detectable Analytes BKV
Type BKV Control
Level High
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.