Learn More
Thermo Scientific™ AcroMetrix™ HBV Controls
Catalog No. p-7000132
Click to view available options
Level:
High
Low
Mid
Identify increases in random or systematic error of nucleic acid tests for hepatitis B viral DNA (HBV DNA) with these controls, which are for In Vitro Diagnostic Use.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of hepatitis B viral DNA (HBV DNA) with the Thermo Scientific™ AcroMetrix™ HBV Controls. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HBV Low Control
Detectable Analytes HBV
Type HBV Control
Level Low
Packaging 5 x 1mL
Quantity 5 x 1 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.