Learn More
Thermo Scientific™ AcroMetrix™ HIV-1 Controls

Description
Assess the performance of molecular diagnostic test procedures, including the RNA extraction step, for the quantitative and qualitative determination of human immunodeficiency virus type-1 (HIV-1) RNA with the Thermo Scientific™ AcroMetrix™ HIV-1 Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Description | AcroMetrix HIV-1 Low Control |
Detectable Analytes | HIV-1 |
Type | HIV-1 Control |
Packaging | 5 x 1.2mL |
Quantity | 5 x 1.2 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.