Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD)
Description
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis C and Hepatitis B in patient samples with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD). Provided in 50mL bulk format and composed of multiple analytes, the AcroMetrix HIV-1/HCV/HBV Controls provide end-user workflow flexibility and cost-efficiency. Manufactured in accordance with ISO 17511, these controls are designed to yield consistent results from lot-to-lot and are ideal for monitoring assay performance over time. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Detectable Analytes | HIV-1, HCV, HBV |
Type | HIV-1/HCV/HBV Control |
Level | High |
Packaging | 50mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.