Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD)
GSA_VA
Catalog No. 23279128
Change view
Click to view available options
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-128 Low
23-279-129 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-279-128 Supplier Thermo Scientific™ Supplier No. 967150
Add to Cart
Add to Cart
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis B and C with bulk controls, which are for In Vitro Diagnostic Use.
Assess the performance of molecular assays that detect human immunodeficiency virus type 1, Hepatitis C and Hepatitis B in patient samples with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Controls (IVD). Provided in 50mL bulk format and composed of multiple analytes, the AcroMetrix HIV-1/HCV/HBV Controls provide end-user workflow flexibility and cost-efficiency. Manufactured in accordance with ISO 17511, these controls are designed to yield consistent results from lot-to-lot and are ideal for monitoring assay performance over time. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes HIV, HCV, HBV
Type HIV-1/HCV/HBV Control
Level Low
Packaging 50mL