Learn More
Thermo Scientific™ AcroMetrix™ MRSA Positive Control
Description
Assess the performance of quantitative and qualitative molecular test procedures for the determination of MRSA DNA with the Thermo Scientific™ AcroMetrix™ MRSA Positive Control. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The control is for In Vitro Diagnostic Use.
Specifications
Specifications
| Type | MRSA Control |
| Packaging | 5 x 0.2mL |
| Quantity | 5 x 0.2 mL |