Learn More
Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO)

Description
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | AcroMetrix PeliSpy Sero Type 36 Control |
| Detectable Analytes | anti-HBc, anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg |
| Sample Type | Serum |
| Type | Control |
| Packaging | 10 x 5mL |
| Quantity | 10 x 5 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.