Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ AcroMetrix™ Controls (BB NAT)
Click to view available options
Description:
AcroMetrix BB NAT HBV Positive Control
AcroMetrix BB NAT HCV Positive Control
AcroMetrix BB NAT HIV-1 Positive Control
AcroMetrix BB NAT Negative Control
Packaging:
10 x 1.4mL
Description
Assess the analytical performance of molecular diagnostic test procedures for the qualitative determination of viral nucleic acids with the Thermo Scientific™ AcroMetrix™ Controls. These controls are intended for use with the detection of viral nucleic acids from donations of whole blood and blood components for transfusion. The controls are for In Vitro Diagnostic Use.

Specifications
Specifications
Description | AcroMetrix BB NAT HBV Positive Control |
Detectable Analytes | HBV |
Sample Type | Plasma, Serum |
Type | Control |
Packaging | 10 x 1.4mL |
Quantity | 10 x 1.4 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction