Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ Controls (BB NAT)

Catalog No. 23279176
Click to view available options
Description:
AcroMetrix BB NAT HBV Positive Control
AcroMetrix BB NAT HCV Positive Control
AcroMetrix BB NAT HIV-1 Positive Control
AcroMetrix BB NAT Negative Control
Packaging:
10 x 1.4mL

Assess the analytical performance of NAT test procedures for the qualitative determination of viral nucleic acids with these controls, which are for In Vitro Diagnostic Use.

Assess the analytical performance of molecular diagnostic test procedures for the qualitative determination of viral nucleic acids with the Thermo Scientific™ AcroMetrix™ Controls. These controls are intended for use with the detection of viral nucleic acids from donations of whole blood and blood components for transfusion. The controls are for In Vitro Diagnostic Use.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix BB NAT Negative Control
Detectable Analytes Negative Control
Sample Type Plasma, Serum
Type Control
Packaging 10 x 1.4mL
Quantity 10 x 1.4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.