Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO)
GSA_VA

Catalog No. p-7000129
Click to view available options
Description:
AcroMetrix Adenovirus Plasma Panel
AcroMetrix BKV Panel
AcroMetrix CMVtc Panel
AcroMetrix EBV Plasma Panel
AcroMetrix HSV-1 Plasma Panel
AcroMetrix HSV-2 Plasma Panel
AcroMetrix VZV Plasma Panel
Packaging:
5 x 0.5mL
5 x 0.8mL
Level:
100, 1000, 10000, 100000, 1000000 IU/mL
100, 1000, 10000, 100000, 1000000 copies/mL
1000, 10000, 100000, 1000000, 10000000 copies/mL
300, 3000, 30000, 300000, 3000000 IU/mL
500, 5000, 50000, 500000, 5000000 copies/mL

Assess the analytical performance of molecular test procedures for transplant viruses with these panels, which are for Research Use Only.

Assess the analytical performance of molecular test procedures, including the nucleic acid extraction step, for the quantitative and qualitative determination of viruses related to transplant disease with the Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits, and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix VZV Plasma Panel
Type Plasma Panel
Level 100, 1000, 10000, 100000, 1000000 copies/mL
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.