Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrosin Antibody (ACR-2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500419IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500419IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500419IR Supplier Novus Biologicals Supplier No. NB500419IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acrosin Monoclonal specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Acrosin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone ACR-2
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Gene Alias acrosin, ACRS, preproacrosin, proacrosin
Gene Symbols ACR
Host Species Mouse
Immunogen Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 49
Test Specificity The antibody ACR-2 reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa.
Target Species Pig, Bovine (Negative), Canine (Negative), Human (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.