Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACRV1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ACRV1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9170
![]() ![]() |
Proteintech Group Inc
140401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
One of the most frequent causes of man infertility is defective sperm acrosome. This damage can be detected using antibodies against intra-acrosomal proteins. Besides diagnostics of sperm pathology, monoclonal antibodies against intra-acrosomal proteins can be used for evaluation of the physiological state of sperm cells as well as for selection of a suitable method of fertilization in the laboratories of assisted reproduction.Specifications
ACRV1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P26436 | |
56 | |
ACRV1 Fusion Protein Ag5109 | |
Primary | |
-20°C | |
ACRV1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Acrosomal protein SP 10, acrosomal vesicle protein 1, ACRV1, D11S4365, SP 10, SPACA2 | |
ACRV1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title