Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Acrylic Equilateral Prism
SDPSCI_ED

Catalog No. S15818 Shop All United Scientific Supplies, Inc Products
Click to view available options
Length (Metric):
25 mm
50 mm
75 mm
100 mm
125 mm
150 mm

Ideal for classroom use.

  • This equilateral prism is ideal for lessons on optics, wavelengths and more.
  • Made from quality clear acrylic sheet with highly polished surfaces
  • Available in six sizes

Specifications

Material Acrylic
Product Type Prism
For Use With (Application) Optics and Light
Length (Metric) 125 mm
Length (English) 4.9 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.