Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSBG1 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
SDP

Catalog No. p-200010635
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2472 30 μL
50-167-2329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2472 Supplier OriGene Supplier No. TA501187S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSBG1 Monoclonal antibody specifically detects ACSBG1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ACSBG1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D8
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DDX39B
Gene Accession No. Q13838
Gene Alias ACSBG1; Acyl-CoA synthetase bubblegum family member 1; BG; BG1; BG1 protein; BGM; bubblegum; E230019G03Rik; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; Grlacs; GR-LACS; hBG1; KIAA0631; Lipidosin; lipidosis-related protein lipidosin; lipodisin; long-chain-fatty-acid--CoA ligase ACSBG1; LPD; mBG1; mKIAA0631; R75185; very long-chain acyl-CoA synthetase
Host Species Mouse
Immunogen Full length human recombinant protein of human BAT1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7919
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.