Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB11099585W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11099585W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11099585W Supplier Novus Biologicals Supplier No. NB11099585AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSL1 Polyclonal specifically detects ACSL1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACSL1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P33121
Gene Alias ACS1LACS 2, Acyl-CoA synthetase 1, acyl-CoA synthetase long-chain family member 1, EC 6.2.1, FACL1EC 6.2.1.3, fatty-acid-Coenzyme A ligase, long-chain 1, LACS 1, LACSlong-chain 2, lignoceroyl-CoA synthase, Long-chain acyl-CoA synthetase 1, Long-chain acyl-CoA synthetase 2, Long-chain fatty acid-CoA ligase 2, long-chain fatty-acid-coenzyme A ligase 1, long-chain-fatty-acid--CoA ligase 1, Palmitoyl-CoA ligase 1, Palmitoyl-CoA ligase 2, paltimoyl-CoA ligase 1
Gene Symbols ACSL1
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human ACSL1 protein (within residues 1-100). [Swiss-Prot# P33121]
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2180
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.