Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11099585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11099585 0.1 mL
NB11099585T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11099585 Supplier Novus Biologicals Supplier No. NB11099585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ACSL1 Polyclonal specifically detects ACSL1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACSL1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:25 - 1:100
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P33121
Gene Alias ACS1LACS 2, Acyl-CoA synthetase 1, acyl-CoA synthetase long-chain family member 1, EC 6.2.1, FACL1EC 6.2.1.3, fatty-acid-Coenzyme A ligase, long-chain 1, LACS 1, LACSlong-chain 2, lignoceroyl-CoA synthase, Long-chain acyl-CoA synthetase 1, Long-chain acyl-CoA synthetase 2, Long-chain fatty acid-CoA ligase 2, long-chain fatty-acid-coenzyme A ligase 1, long-chain-fatty-acid--CoA ligase 1, Palmitoyl-CoA ligase 1, Palmitoyl-CoA ligase 2, paltimoyl-CoA ligase 1
Gene Symbols ACSL1
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human ACSL1 protein (within residues 1-100). [Swiss-Prot# P33121]
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2180
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.