Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB060128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060128 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060128 Supplier Novus Biologicals Supplier No. NBP299339FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSL1 Polyclonal specifically detects ACSL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ACSL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ACS1LACS 2, Acyl-CoA synthetase 1, acyl-CoA synthetase long-chain family member 1, EC 6.2.1, FACL1EC 6.2.1.3, fatty-acid-Coenzyme A ligase, long-chain 1, LACS 1, LACSlong-chain 2, lignoceroyl-CoA synthase, Long-chain acyl-CoA synthetase 1, Long-chain acyl-CoA synthetase 2, Long-chain fatty acid-CoA ligase 2, long-chain fatty-acid-coenzyme A ligase 1, long-chain-fatty-acid--CoA ligase 1, Palmitoyl-CoA ligase 1, Palmitoyl-CoA ligase 2, paltimoyl-CoA ligase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ACSL1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2180
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.