Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL5 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB330337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330337 Supplier Novus Biologicals Supplier No. NBP297725PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACSL5 Polyclonal antibody specifically detects ACSL5 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ACSL5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ACS2, ACS5FACL5 for fatty acid coenzyme A ligase 5, acyl-CoA synthetase long-chain family member 5, EC 6.2.1, EC 6.2.1.3, FACL5fatty acid coenzyme A ligase 5, fatty-acid-Coenzyme A ligase, long-chain 5, LACS 5, Long-chain acyl-CoA synthetase 5, long-chain fatty acid coenzyme A ligase 5, long-chain-fatty-acid--CoA ligase 5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ACSL5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51703
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.