Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSM5 Antibody (OTI1G1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271978G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271978G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271978G Supplier Novus Biologicals Supplier No. NBP271978G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACSM5 Monoclonal specifically detects ACSM5 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACSM5
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1G1
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias acyl-CoA synthetase medium-chain family member 5, acyl-coenzyme A synthetase ACSM5, mitochondrial, EC 6.2.1, EC 6.2.1.2, FLJ20581, MACS3
Gene Symbols ACSM5
Host Species Mouse
Immunogen Full length recombinant protein of human ACSM5 (NP_060358) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54988.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.