Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACT16 Antibody (GB110A), Novus Biologicals™
SDP

Catalog No. NBP261092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261092 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261092 Supplier Novus Biologicals Supplier No. NBP261092
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACT16 Monoclonal specifically detects ACT16 in Bovine, Water Buffalo samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen ACT16
Applications Flow Cytometry, Immunohistochemistry, Functional Assay
Classification Monoclonal
Clone GB110A
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Ascites with 0.09% Sodium Azide
Host Species Mouse
Immunogen Goat ConA stimulated mononuclear leukocytes
Purification Method Unpurified
Quantity 0.1 mg
Primary or Secondary Primary
Target Species Water Buffalo
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.