Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTH Antibody (CLIP/2859R), Alexa Fluor™ 405 - C-terminus, Novus Biologicals™
SDP

Catalog No. NB385445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385445 Supplier Novus Biologicals Supplier No. NBP308633AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ACTH Monoclonal antibody specifically detects ACTH in Human samples. It is validated for ELISA

Specifications

Antigen ACTH
Applications ELISA
Classification Monoclonal
Clone CLIP/2859R
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) (Uniprot: P01189)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5443
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.