Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTH, Mouse anti-Human, Clone: CLIP/1418, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280544100UG
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA.Specifications
| ACTH | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 1:100-1:2000 | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5443 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA | |
| CLIP/1418 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| POMC | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction