Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTH, Mouse anti-Human, Clone: CLIP/1418, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | ACTH |
---|---|
Clone | CLIP/1418 |
Concentration | 0.2 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA.Specifications
ACTH | |
0.2 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
POMC | |
IgG1 κ | |
Protein A or G purified |
CLIP/1418 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
5443 | |
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title