Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Antibody (3B3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP197723UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197723UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197723UV Supplier Novus Biologicals Supplier No. NBP197723UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin Monoclonal specifically detects Actin in Human, Rat, Porcine, Goat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen Actin Alpha 1 Skeletal Muscle
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone 3B3
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Gene Symbols ACTA1
Host Species Mouse
Immunogen Clone 3B3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-skeletal actin with an acetylated N-terminus (Ac-DEDETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Test Specificity Clone 3B3 is an antibody highly specific for alpha-skeletal actin, and does not cross react with other actin isoforms.
Target Species Human, Rat, Pig, Goat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.