Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Antibody (9Y9L6), Novus Biologicals™
SDP

Catalog No. NB168858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB168858 Supplier Novus Biologicals Supplier No. NBP316885
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin Monoclonal antibody specifically detects Actin in Plant samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Actin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 9Y9L6
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 1% BSA, 50% glycerol, pH7.3
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Host Species Mouse
Immunogen Recombinant Protein of Actin (plant specific)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Plant
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.