Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Actin Cytoskeleton / Focal Adhesion Staining Kit

Catalog No. fak100 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 Assays

Includes

  • Vinculin Monoclonal antibody, purified clone 7F9. One vial containing 100μL at 1mg/mL
  • TRITC-conjugated Phalloidin. One vial containing 15μg
  • DAPI. One vial containing 100μL

Very sensitive immunocytochemical tool that contains fluorescent-labeled phalloidin (TRITC-conjugated phalloidin) to map the local orientation of actin filaments within cell and mouse monoclonal antibody to Vinculin that is very specific for the staining of focal contacts in cells

  • Kit consists of three components (TRITC-conjugated phalloidin, anti-Vinculin and DAPI) for the immunofluorescent staining of actin filaments in the cytoskeleton, focal contacts as well as the nucleus of the cells
  • Reagents and materials supplied in the Actin Cytoskeleton and Focal Adhesion Staining Kit are sufficient for 100 tests (including controls)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage When stored at 2° to 8°C, the kit components are stable up to the expiration date
For Use With (Application) Immunocytochemistry
Quantity 100 Assays
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.