Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Gamma 1 Antibody (2A3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP197720IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197720IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197720IR Supplier Novus Biologicals Supplier No. NBP197720IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Porcine, Chicken, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen Actin Gamma 1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone 2A3
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Gene Accession No. P63261
Gene Alias ACT, ACTB, ACTG, actin, cytoplasmic 2, actin, gamma 1, cytoskeletal gamma-actin, deafness, autosomal dominant 20; deafness, autosomal dominant 26, DFNA20, DFNA26, Gamma-actin
Gene Symbols ACTG1
Host Species Mouse
Immunogen 2A3 is a Mouse monoclonal IgG2b antibody derived by fusion of NS0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a peptide comprising the N-terminal nonapeptide of gamma-cytoplasmic actin with an acetylated N-terminus (Ac-EEEIAAL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 71
Test Specificity 2A3 is highly specific for gamma-cytoplasmic actin (gamma-actin 1; encoded by ACTG1). It does not cross-react with the gamma-actin isoform expressed in the enteric smooth muscle tissues (gamma-actin 2; encoded by ACTG2) and does not cross react with other
Target Species Human, Mouse, Rat, Pig, Chicken, Mammalia, Rabbit, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.