Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Mouse anti-Human,Eukaryotes, Unconjugated, Clone: MAb GEa (5-16 H1C8), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT1533100UL
Description
Actin Monoclonal antibody specifically detects Actin in Human,Eukaryotes samples. It is validated for ELISA, Western Blotting.
Specifications
Actin | |
Monoclonal | |
Unconjugated | |
ACT1 | |
Mouse | |
∼44 kDa observed; 41.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μL | |
Primary | |
Human, Eukaryote | |
Purified |
ELISA, Western Blot | |
MAb GEa (5-16 H1C8) | |
NP_850284.1 | |
ACT1, AAC1, At2g37620, E13M22.12 | |
Purified actin (ACT1) from Arabidopsis thaliana plant. | |
Ligatrap | |
Cell Structure | |
Clone 5-16 H1C8 is a mouse monoclonal antibody that detects Actin in a wide range of species. | |
Stable for 1 year at -20°C from date of receipt. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction