Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Mouse anti-Human,Eukaryotes, Unconjugated, Clone: MAb GEa (5-16 H1C8), MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | Actin |
---|---|
Clone | MAb GEa (5-16 H1C8) |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15332MI
|
MilliporeSigma
MABT153325UL |
25 μL |
Each for $189.29
|
|
|||||
MABT15331MI
|
MilliporeSigma
MABT1533100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
Actin Monoclonal antibody specifically detects Actin in Human,Eukaryotes samples. It is validated for ELISA, Western Blotting.Specifications
Actin | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human, Eukaryote | |
ACT1 | |
Purified actin (ACT1) from Arabidopsis thaliana plant. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼44 kDa observed; 41.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
MAb GEa (5-16 H1C8) | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_850284.1 | |
ACT1, AAC1, At2g37620, E13M22.12 | |
IgM | |
Ligatrap | |
Clone 5-16 H1C8 is a mouse monoclonal antibody that detects Actin in a wide range of species. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title