Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin (Muscle Specific) Antibody (MSA/953), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N24762PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N24762PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N24762PE594 Supplier Novus Biologicals Supplier No. NBP247662PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin (Muscle Specific) Monoclonal specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Flow Cytometry.

Specifications

Antigen Actin (Muscle Specific)
Applications Flow Cytometry
Classification Monoclonal
Clone MSA/953
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P62736, P68032, P68133
Gene Alias ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3
Gene Symbols ACTA1
Host Species Mouse
Immunogen Synthetic peptide common among human muscle actins
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58.0
Test Specificity This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium. Actin can be resolved on the basis of its isoelectric points into three distinctive components: alpha, beta, and gamma in order of increasing isoelectric point. Anti-muscle specific actin recognizes alpha and gamma isotypes of all muscle groups. Non-muscle cells such as vascular endothelial cells and connective tissues are non-reactive. Also, neoplastic cells of non-muscle-derived tissue such as carcinomas, melanomas, and lymphomas are negative. It stains tumors of smooth muscle (leiomyomas and leiomyosarcomas) as well as skeletal muscle (rhabdomyomas and rhabdomyosarcomas).
Target Species Human, Rat, Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.