Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin (Muscle Specific) Antibody (rMSA/953), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB179603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179603 Supplier Novus Biologicals Supplier No. NBP314096V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Actin (Muscle Specific)
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMSA/953
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3
Host Species Mouse
Immunogen Recombinant protein corresponding to human muscle actin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Human, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.