Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin1 Antibody (mAb45a), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP253123V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP253123V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP253123V Supplier Novus Biologicals Supplier No. NBP253123V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin1 Monoclonal specifically detects Actin1 in Plant samples. It is validated for Western Blot.

Specifications

Antigen Actin1
Applications Western Blot
Classification Monoclonal
Clone mAb45a
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AAC1, Actin-1
Host Species Mouse
Immunogen Purified Arabidopsis Actin1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Signaling
Primary or Secondary Primary
Test Specificity Recognizes subclasses 4 and 5 of vegetative plant actins (arabidopsis ACT1, 3, 4, 12).
Target Species Plant
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.