Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin2 Antibody (mAb13a), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP253122R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP253122R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP253122R Supplier Novus Biologicals Supplier No. NBP253122R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin2 Monoclonal specifically detects Actin2 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Actin2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone mAb13a
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACT2, actin 2, Actin-2
Host Species Mouse
Immunogen Purified Arabidopsis Actin2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Signaling
Primary or Secondary Primary
Test Specificity This mouse monoclonal antibody was generated against plant actin ACT2 and recognizes subclasses 1 and 3 of vegetative plant actins (arabidopsis ACT2, 8, 11).
Target Species Plant
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.